Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Solus™ 2000 Series Protective Eyewear
Comfortable, stylish eyewear with a vast array of color and lens tint options. Eye protection with a stylish flare.
Supplier: 3M™ 7100203151
This item is not returnable.
View return policy
Description
- Adjustable ratchet temples
- Polycarbonate lenses absorb 99.9% of UVA and UVB rays
- Meet the requirements of ANSI Z87.1-2015 Standard as an Impact Rated Spectacle
- 3M Catalog# S2001AF-BLU, S2001AS-BLU, S2001SGAF-BGR, S2001SGAF-BGR-F, S2001SGAF-RED, S2002SGAF-BGR, S2002SGAF-RED, S2003SGAF-BGR, S2005SGAF-BGR, S2007SGAF-BGR, S2017AS-BLK, S2024AS-RED, S2030AS-BLK, S2050AS-BLK
Specifications
Anti-fog, Scratch-resistant | |
Protective eyewear | |
Blue | |
Dual Injected Temple | |
Anti-Scratch, Anti-Fog | |
Plastic |
Clear | |
Assembly, Law Enforcement, Primary Metals, Power Fastening, Pouring/Casting | |
S2001AF-BLU | |
Blue, Gray | |
Polycarbonate | |
0.999 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title