Learn More
3M™ Solus™ 2000 Series Safety Glasses
3M™ Solus™ 2000 Series Safety Glasses meet ANSI Z87.1-2015 Standards for an Impact Rated Spectacle
$283.20
Specifications
Certifications/Compliance ANSI Z87.1-2015
For Use With (Application) Chemical Handling, Machining, Oil and Gas, Automotive, Construction, Primary Metals, Law Enforcement, Mining, Power Fastening, Chiseling, Drilling, Pouring and Casting, Painting, Grinding, Food and Beverage Manufacturing, Masonary, Remediation, Sub Applications, Demolition, Food and Beverage, Aviation, Machine Operations, Assembly, Pharmaceuticals, Facility Maintenance, General Manufacturing, Chemicals
Frame Color Clear
Frame Material Plastic
Product Type Safety glasses
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
19-072-208
View Documents
3M™
7100221706
Model: S2008AS-CLR, Scratch-resistant, Lens Tint: Blue/Mirror
Case of 20 for $283.20
Add to cart
19-072-210
View Documents
3M™
7100221705
Model: S2001SGAF-CLR, Anti-Fog/Scratch-resistant, Lens Tint: Clear
Case of 20 for $283.20
Add to cart
Description
Description
  • Solus™ 2000 Series provides eye protection with a stylish flare
  • Adjustable ratchet temples allow the wearer to customize their desired fit
  • Optional 3M™ Scotchgard™ Anti-Fog lens coating resists fogging even after up to 25 washings
  • Polycarbonate lenses absorb 99.9% of UVA and UVB rays
Specifications
Specifications
ANSI Z87.1-2015
Clear
Safety glasses
Dual Injected Temple
Chemical Handling, Machining, Oil and Gas, Automotive, Construction, Primary Metals, Law Enforcement, Mining, Power Fastening, Chiseling, Drilling, Pouring and Casting, Painting, Grinding, Food and Beverage Manufacturing, Masonary, Remediation, Sub Applications, Demolition, Food and Beverage, Aviation, Machine Operations, Assembly, Pharmaceuticals, Facility Maintenance, General Manufacturing, Chemicals
Plastic
Clear, Blue
0.999