Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
solute carrier family 22, member 18 antisense Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP215120
Description
solute carrier family 22, member 18 antisense Polyclonal specifically detects solute carrier family 22, member 18 antisense in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
solute carrier family 22, member 18 antisense | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
SLC22A18AS | |
Recombinant protein encompassing a sequence within the center region of human SLC22A18AS. The exact sequence is proprietary. | |
0.1 mL | |
Primary | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.75mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
BWR1BSolute carrier family 22 member 18 antisense protein, BWSCR1BBeckwith-Wiedemann region 1B, ORCTL2Sbeckwith-Wiedemann syndrome chromosomal region 1 candidate gene B protein, organic cation transporter-like 2 antisense, Organic cation transporter-like protein 2 antisense protein, p27-BWR1Bp27-Beckwith-Wiedemann region 1 B, SLC22A1LSBeckwith-Wiedemann syndrome chromosome region 1, candidate b, solute carrier family 22 (organic cation transporter), member 18 antisense, solute carrier family 22 (organic cation transporter), member 1-like antisense, Solute carrier family 22 member 1-like antisense protein | |
Rabbit | |
Affinity Purified | |
RUO | |
5003 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction