Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Solution A For USP Testing of Atropine Sulfate, Reagents
Solution A For USP Testing of Atropine Sulfate, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CA0166001C
Specifications
Solution A For USP Testing of Atropine Sulfate | |
7732-18-5, 7790-98-9, 7790-98-9 | |
USP | |
1.0094 | |
1 L | |
Liquid |
Colorless | |
100°C | |
Glass Bottle | |
3 | |
Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction