Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Solution D
Solution D
$197.65 - $1959.65
Specifications
Content And Storage | Store in freezer (-5 to -30°C). |
---|---|
Shipping Condition | Dry Ice |
Product Line | GeneBLAzer™ |
For Use With (Application) | Pharma & Biopharma; Protein Assays and Analysis; Cellular Imaging |
Description
Solution D is an anion transport inhibitor, for cells such as CHO-K1, that can be used in conjunction with CCF2-AM and CCF4-AM-based studies to prevent active cellular export of the FRET-based substrates after loading.Specifications
Store in freezer (-5 to -30°C). | |
Dry Ice | |
GeneBLAzer™ | |
Pharma & Biopharma; Protein Assays and Analysis; Cellular Imaging |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title