Learn More
Somatostatin Mouse anti-Human, Clone: U24-354, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566031
Catalog No. BDB566031
Add to cart
Description
Description
Somatostatin Monoclonal antibody specifically detects Somatostatin in Human, Mouse, Rat samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications
Specifications
Somatostatin
U24-354
Unconjugated
Purified
Mouse
Mouse IgG2b, κ
RUO
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Monoclonal
Flow Cytometry, Flow Cytometry (Intracellular Staining)
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
growth hormone release-inhibiting factor, SST
Human Preprosomatostatin Recombinant Protein
0.1mg
Cell Biology
Primary
Human, Murine, Rat
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.