Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Somatostatin R3/SSTR3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | Somatostatin R3/SSTR3 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB124965
|
Novus Biologicals
NBP310032100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
Somatostatin R3/SSTR3 Polyclonal specifically detects Somatostatin R3/SSTR3 in Human samples. It is validated for Western Blot.Specifications
Somatostatin R3/SSTR3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Breast Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS buffer, 2% sucrose | |
6753 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
somatostatin receptor 3, SS-3-R, SS3-R | |
The immunogen is a synthetic peptide directed towards the C terminal region of human Somatostatin R3/SSTR3 (NP_001042.1). Peptide sequence GGKGKEMNGRVSQITQPGTSGQERPPSRVASKEQQLLPQEASTGEKSSTM | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title