Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Somatostatin R5/SSTR5, Rabbit anti-Human, Alexa Fluor 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB10056498AF405

Catalog No. NB003713


Add to cart

Description

Description

Somatostatin R5/SSTR5 Polyclonal specifically detects Somatostatin R5/SSTR5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Somatostatin R5/SSTR5
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
somatostatin receptor 5, somatostatin receptor subtype 5, somatostatin receptor type 5, SS5R, SS-5-R, SS5-R
Rabbit
IgG
0.1 mL
Cell Cycle and Replication, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
The Somatostatin Receptor 5 amino acid sequence used as immunogen for this antibody is 100% homologous in human and 94% in chimp. The expected size of SSRT5, based on amino acid content, is approximately 40 kD. It is believed that the 5-10 kD increase in observed size is due to glycosylation.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Affinity Purified, Conjugated
SSTR5
A portion of amino acids 300-350 of human SSTR5 was used as the immunogen.
Immunogen affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit