Learn More
SOMNI Scientific™ SOMNI™ 19.1 Cagemount Vaporizer
Incorporates original design with updated functionality, advanced wicking and 20% added capacity. The one-piece redesigned filling system reduces spillage and unnecessary agent exposure. The ideal vaporizer for extended preclinical surgical procedures.
Supplier: SOMNI Scientific™ VS6008
Description
- Quality materials
- Brass, stainless steel
- Advanced wicking system
- Remodeled original design with updated functionality
- Calibrated using laser refractometry assures accuracy
- Safe, leak proof performance
- Advanced wicking system assures linear vapor delivery
- One piece fill system
- 2 year service cycle
Specifications
Animal Inhalant Anesthesia | |
Brass and Alloys | |
15.2 cm | |
Mounting hardware, Calibration Certificate |
RM-17003, RM-17004, AMD-17004, TT-17000, OC-21003, OC-21004 | |
19.4 cm | |
Isoflurane KeyFill |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Veterinary Use Only