Learn More
SOMNI Scientific™ SOMNI Scientific™ Rebreathing Bag
SDP
Catalog No. 04777200 Shop All Somni Scientific Products
Click to view available options
Capacity (Metric):
0.5 L
1 L
2 L
3 L
The bag is designed to inflate uniformly and has a reinforced neck. Latex, PVC and BPA free
  • The rebreathing bag is designed to inflate uniformly and has a reinforced neck
  • Latex free
  • PVC and BPA free
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
Type Breathing Bag
Capacity (Metric) 0.5 L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.