Learn More
SOMNI Scientific™ SOMNI Scientific™ SOMNI Bain Circuit
SDP
The tube within a tube design provides heat and moisture conservation with fresh gas entering the circuit at the patient end. GS elbow included. PVC, Phthalate and Latex Free
Supplier:  SOMNI Scientific™ BC13020
 View more versions of this product
Catalog No. 04-777-212
Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative.
Description
Description
  • A tube within a tube design that provides heat and moisture conservation
  • The fresh gas enters the circuit at the patient end
  • PVC, Phthalate and Latex Free
  • GS elbow included
Specifications
Specifications
Research Animal Inhalant Anesthesia
120 in.