Learn More
SOMNI Scientific™ SOMNI Scientific™ SOMNI Bain Circuit
The tube within a tube design provides heat and moisture conservation with fresh gas entering the circuit at the patient end. GS elbow included. PVC, Phthalate and Latex Free
$45.60 - $68.40
Specifications
For Use With (Application) Research Animal Inhalant Anesthesia
View More Specs
Products 3
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
04-777-210
View Documents
SOMNI Scientific™
BC13001
72 in.
Each for $45.60
Add to cart
04-777-211
View Documents
SOMNI Scientific™
BC13011
60 in.
Each for $48.90
Add to cart
04-777-212
View Documents
SOMNI Scientific™
BC13020
120 in.
Each for $68.40
Add to cart
Description
Description
  • A tube within a tube design that provides heat and moisture conservation
  • The fresh gas enters the circuit at the patient end
  • PVC, Phthalate and Latex Free
  • GS elbow included
Specifications
Specifications
Research Animal Inhalant Anesthesia