Sonoco™ ThermoSafe Certis™ Silver Shipper Box with Summer Packout
Accommodates broad range of temperature-sensitive products
Manufacturer: Sonoco™ CRSS3SE
 View all options for this product
Catalog No. 03-531-215
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Material Expanded Polystyrene (EPS)
    Color Silver
    Elapsed Time Range 78hrs.
    Nominal Payload (English) 59.1 lb.
    With flexible, modular design, these shippers can adjust to meet customer requirements for temperature control, shipping durations and payload sizes. ThermoPhase™ powered by PureTemp™ non-petroleum phase-change technology enables strict temperature control, easy pack outs, greater efficiency and packages that are smaller and lighter. Three packout options include seasonal and year-round temperature protection.
    • Choice of two ambient profiles (ISCgold™ and ISCsilver™)
    • Easily modified to align with product specifications
    • Off-the-shelf convenience
    • Reduced shipping costs and transit time to market
    Certis Silver:
    • ISCsilver ambient profile to protect packages under moderate or temperature-controlled distribution conditions
    • One to three day durations
    • Seasonal and Universal packouts
  • Description & Specifications
Related Products
Product Content Feedback