Learn More
Sonoco™ ThermoSafe DiagnosticShipper Systems, Without Tubes Available on GSA/VA Contract for Federal Government customers only.
Ready-to-use combination/triple packaging kits
Manufacturer:  Sonoco™ 570
 View more versions of this product
Catalog No. 03-52-556
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Absorbs maximum liquid capacity
Conforms with DOT, USPS, IATA packing instruction 650
Plastic envelope, absorbent material, bubble wrap
Diagnostic specimen shipping system without tubes
Accessories