Learn More
Sonoco™ ThermoSafe EPS Foam and Corrugate Shippers Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Sonoco™ 658
 View more versions of this product
Catalog No. 03-001-237
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Safe storage and delivery of refrigerated or frozen material (pharmaceuticals, laboratory specimens, serums, vaccines, human organs, culture pack and nuclear and isotope materials)
<24 hr.
10.875 in.
21.6cm
15.8 in.
3 lb.
15.12 in.
91kg test corrugated carton
Rounded corners and edges
8.5 in.
18.12 in.
Molded Expanded Polystyrene
1.4kg
12.8 in.