Learn More
Sonoco™ ThermoSafe Infectious Shipper Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Sonoco™ INF3000
 View more versions of this product
Catalog No. 03-600-149
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
DOT Regulation 49 CFR Parts 171 to 178 and IATA Packing Instructions 602
Up to 50mL
7.5 in.
Compartmentalized bubble pouch
6.25 x 6.25 x 7.5 in. (15.8 x 15.8 x 19cm)
19cm