Learn More
Sonoco™ ThermoSafe Infectious Shipper Overpack
Infectious substance shipper overpack
Manufacturer:  Sonoco™ 670
Catalog No. 11-675-97
Add to cart
Description
Description
  • EPS foam container and lid
  • Accommodates Model 630 or Model 660B shipper plus 25 lb. of dry ice
  • Foam insert holds in place while dry ice sublimates
  • Includes one Class 6 Infectious Substance label and one Class 9 Miscellaneous Dangerous Goods label (for dry ice) and shipping carton
Specifications
Specifications
Oone Class 6 Infectious Substance label and one Class 9 Miscellaneous Dangerous Goods label (for dry ice) and shipping carton
Expanded Polystyrene Foam
Documents
Accessories