Learn More
Sonoco™ ThermoSafe InsulatedShipper-PUR, Packed in Case Available on GSA/VA Contract for Federal Government customers only.
High performance polyurethane containers meet the needs of the most demanding temperature sensitive payloads
Manufacturer:  Sonoco™ E9UPS
 View more versions of this product
Catalog No. 03-532-155
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Polyurethane
8 in.
20.3cm
4.5 in.
11.4cm
4.5 in.
11.4cm
14.12 in.
35.8cm
9.5 in.
24.1cm
9.12 in.
23.1cm
9.5 x 9.12 x 14.12 in. (24.1 x 23.1 x 35.8cm)