Sonoco™ ThermoSafe LabMailer Sample-Guard 28mL Vial Shippers
Insulated shippers with lid eliminate sample leakage and breakage
Manufacturer: Sonoco™ 480490
 View more versions of this product
Catalog No. 11-675-47
  Check Availability
Add to Cart
    Specifications
    Material Insulated Expanded Polystyrene (EPS) Foam
    Includes One 24 oz. Phase Change Material Reusable Pack
    Quantity 48 x 40mL Vials
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback