Learn More
Sonoco™ ThermoSafe LabMailer for Vials
Lightweight, economical EPS shippers provide maximum breakage protection for known nonhazardous materials
Manufacturer:  Sonoco™ 490C24
Catalog No. 11-676-26A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
A unique array of shippers fills the need for protection of frozen, refrigerated, and ambient specimens required for compliance, analysis, testing, research and development, public health, industrial hygiene and environmental use.
  • Accommodates VacuTainer™, Microtainer™, and other collection/transport tubes
  • Available in multi-tube, slide, and tube/slide combinations
  • Lightweight polystyrene foam construction saves postage
  • Reusable and recyclable
  • Foam mailer assembled in corrugated carton
  • Holds 48 vials (16 - 20 mm vials)
Specifications
Specifications
48 Vials (16 - 20 mm vials)
Polystyrene Foam
Documents
Accessories