Learn More
Sonoco™ ThermoSafe Multi-Purpose EPS Foam and Corrugate Shippers Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Sonoco™ 617UPS
 View more versions of this product
Catalog No. 03-530-81
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Molded expanded Polystyrene
7.875 in.
11.73cm
9.5 in.
2.3kg
8.25 in.
91kg Test corrugated carton
<24 hr.
4.62 in.
12 in.
5 lb.
11.25 in.
Safe storage and delivery of refrigerated or frozen material (pharmaceuticals, laboratory specimens, serums, vaccines, human organs, culture pack and nuclear and isotope materials)
Rounded corners and edges
Accessories