Learn More
Sonoco™ ThermoSafe Storage/Transport Chests Under 0.6 Cu. Ft. Available on GSA/VA Contract for Federal Government customers only.
Ideal for dry ice and wet ice storage, short-term frozen storage, field collection, and cryogenic testing
Manufacturer:  Sonoco™ 311
Catalog No. 11-676-15
Add to cart
Description
Description
  • Low-temperature storage chests include 124.7kg (275 lb.) bursting test laminated fiberboard exterior and expanded polystyrene foam interior; with insulated lid
  • Store 15kg (33 lb.) of dry ice
  • Average holding time of 2.6 days
  • Net weight, 1.8kg (4 lb.)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Dolly not included with chests
Specifications
Specifications
Fiberboard Case
0.6 cu. ft.
Fiberboard
Documents
Accessories