Learn More
Sonoco™ ThermoSafe Storage/Transport Chests Under 2.5 Cu. Ft.
Ideal for dry ice and wet ice storage, short-term frozen storage, field collection, and cryogenic testing
Manufacturer:  Sonoco™ 306
Catalog No. 11-676-17
Add to cart
Description
Description
  • Low-temperature storage chests include 124.7kg (275 lb.) bursting test laminated fiberboard exterior and expanded polystyrene foam interior; with insulated lid
  • Store 59kg (130 lb.) of dry ice
  • Average holding time of 11.5 days
  • Net weight, 4.1kg (9 lb.)
Order Info
Dolly not included with chests
Specifications
Specifications
Fiberboard Case
2.5 cu. ft.
Fiberboard