Learn More
Sonoco™ ThermoSafe Storage/Transport Chests Under 4.3 Cu. Ft.
Ideal for dry ice and wet ice storage, short-term frozen storage, field collection, and cryogenic testing
Manufacturer:  Sonoco™ 305
Catalog No. 11-676-18
Add to cart
Description
Description
  • Low-temperature storage chests include 124.7kg (275 lb.) bursting test laminated fiberboard exterior and expanded polystyrene foam interior; with insulated lid
  • Store 122.5kg (270 lb.) of dry ice
  • Average holding time of 18 days
  • Net weight, 6.6kg (14.5 lb.)
Order Info
Dolly not included with chests
Specifications
Specifications
Fiberboard Case
4.3 cu. ft.
Fiberboard
Documents
Accessories