Learn More
Sonoco™ Utek™ Refrigerant Packs
Phase change materials are ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or CO2
Manufacturer:  Sonoco™ 405
 View more versions of this product
Catalog No. 03-528D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
FDA
Ideal for products requiring frozen temperatures or extreme cold that may not tolerate dry ice or Carbon dioxide
Phase Change Material
U-tek™ Mat
16 in. x 34 ft. (40.6cm x 10.3m)
-1°C
1 Case
Accessories