Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Sorbic acid

Click to view available options
Quantity:
100 mg
Description
Sorbic acid is an activator of hFFA2-DREADDs. Inhibits forskolin-induced cAMP production (IC50 ∼ 1 μM) and promotes accumulation of inositol monophosphates (IC50 >1 μM) in cells expressing hFFA2-DREADD. Promotes the release of GLP-1 from colonic crypts derived from hFFA2-DREADD-expressing mice.
Specifications
Specifications
Chemical Name or Material | (2 E,4 E)-2,4-Hexenoic acid |
CAS | 110-44-1 |
Quantity | 100 mg |
Target | DREADD Activators |
Molecular Formula | C6H8O2 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction