Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbic Acid, Powder, FCC, 99-101%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
110-44-1
C6H8O2
112.13
WSWCOQWTEOXDQX-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dienoic acid
C\C=C\C=C\C(O)=O
110-44-1
C6H8O2
112.13
WSWCOQWTEOXDQX-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dienoic acid
C\C=C\C=C\C(O)=O

S1517, 110-44-1, C6H8O2

Spectrum™ Sorbic Acid, Powder, FCC is used as a food preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Ignition Residue 0.002
Water 0.005
Melting Range 132-135°C
Quantity 50 kg
CAS Max % 1
Percent Purity 99 to 101%
Grade FCC