Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbic Acid, Powder, NF, 99-101%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
110-44-1
C6H8O2
112.13
WSWCOQWTEOXDQX-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dienoic acid
C\C=C\C=C\C(O)=O
110-44-1
C6H8O2
112.13
WSWCOQWTEOXDQX-MQQKCMAXSA-N
(2E,4E)-hexa-2,4-dienoic acid
C\C=C\C=C\C(O)=O

SO215, 110-44-1, C6H8O2

Spectrum™ Sorbic Acid, Powder, NF is used as an antimicrobial agent in pharmaceutical formulations. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Ignition Residue 0.002
Water 0.005
Melting Range 132-135°C
Quantity 12 kg
CAS Max % 1
Percent Purity 99 to 101%
Grade NF