Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitan Monolaurate, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
1338-39-2
C18H34O6
346.46
LWZFANDGMFTDAV-IOVMHBDKSA-N
2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl dodecanoate
CCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O
1338-39-2
C18H34O6
346.46
LWZFANDGMFTDAV-IOVMHBDKSA-N
2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl dodecanoate
CCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

SPA20, 1338-39-2, C18H34O6

Spectrum™ Sorbitan Monolaurate , also known as sorbitan laurate, is a mix of esters from the fatty acid lauric acid used as a food additive emulsifier to keep water and oils mixed. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Packaging Poly Drum
Quantity 20 L
Saponification Value 158-175
CAS Max % 1
Grade Reagent