Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitan Monopalmitate, NF, 63-71%, Spectrum™

Click to view available options
Quantity:
100 g
12 kg
2.5 kg
250 lbs.
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
26266-57-9
C22H42O6
402.57
IYFATESGLOUGBX-CBOZIWPYSA-N
2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O
26266-57-9
C22H42O6
402.57
IYFATESGLOUGBX-CBOZIWPYSA-N
2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl hexadecanoate
CCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

SPA43, 26266-57-9, C22H42O6

Spectrum™ Sorbitan Monopalmitate, NF is used as an emulsifier and nonionic surfactant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Ignition Residue 0.005
Water 0.015
Quantity 250 lbs.
Saponification Value 140-150
CAS Max % 1
Percent Purity 63 to 71%
Grade NF