Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitan Monostearate, NF, 68-76%, Spectrum™ Chemical
SDP

Catalog No. p-7158302
Click to view available options
Quantity:
2.5 kg
12 kg
90 kg
Packaging:
Fiber Drum
Poly Pail

SPA63, 1338-41-6, C24H46O6

Spectrum™ Chemical Sorbitan Monostearate, NF is used as an emulsifier and nonionic surfactant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1338-41-6
Molecular Formula C24H46O6
Molecular Weight (g/mol) 430.63
MDL Number MFCD00005366,MFCD00005366
InChI Key HVUMOYIDDBPOLL-IIZJTUPISA-N
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
Ignition Residue 0.005
Packaging Poly Pail
Quantity 2.5 kg
Saponification Value 147-157
Percent Purity 68 to 76%
Grade NF
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.