Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitol, BAKER ANALYZED™ Biochemical Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
3kg
Packaging:
Poly Bottle
Poly Pail
50-70-4
C6H14O6
182.172
FBPFZTCFMRRESA-JGWLITMVSA-N
d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol
5780
CHEBI:17924
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
C(C(C(C(C(CO)O)O)O)O)O
50-70-4
C6H14O6
182.172
FBPFZTCFMRRESA-JGWLITMVSA-N
d-sorbitol,sorbitol,d-glucitol,glucitol,l-gulitol,--sorbitol,diakarmon,glucarine,sorbol,d---sorbitol
5780
CHEBI:17924
(2R,3R,4R,5S)-hexane-1,2,3,4,5,6-hexol
C(C(C(C(C(CO)O)O)O)O)O

High quality biochemicals for research use

Packaging Poly Bottle
Quantity 500g
Specific Gravity 1L = 1.49kg
Formula Weight 182.17
Grade BAKER ANALYZED™ Biochemical Reagent
Chemical Name or Material Sorbitol