Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sorbitol, Powder, NF, 91-100.5%, Spectrum™

Click to view available options
Quantity:
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
50-70-4
C6H14O6
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
hexane-1,2,3,4,5,6-hexol
OCC(O)C(O)C(O)C(O)CO
50-70-4
C6H14O6
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
hexane-1,2,3,4,5,6-hexol
OCC(O)C(O)C(O)C(O)CO

SO219, 50-70-4, C6H14O6

Spectrum™ Sorbitol, Powder, NF is used in the pharmaceutical industry as a sweetener or humectant. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Ignition Residue 0.001
Water 0.015
Quantity 45 kg
pH 3.5 to 7.0
CAS Max % 1
Percent Purity 91 to 100.5%
Grade NF