Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SorCS3 Monoclonal Antibody (339615)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA524071
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant human SorCS1 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications
SorCS3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q8VI51 | |
SORCS3 | |
Mouse myeloma cell line NS0-derived recombinant mouse SorCS3 Glu34-Ser1122. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Immunohistochemistry, Western Blot | |
339615 | |
Unconjugated | |
SORCS3 | |
6330404A12Rik; AW045579; KIAA1059; SORCS; SORCS3; sortilin related VPS10 domain containing receptor 3; sortilin-related VPS10 domain containing receptor 3; VPS10 domain receptor protein SORCS 3 (SORCS3); VPS10 domain-containing receptor SorCS3 | |
Rat | |
Protein A/G | |
RUO | |
66673 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction