Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SORD Mouse anti-Human, Clone: OTI13G5, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA500784

Catalog No. 50-167-1926


Add to cart

Description

Description

Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.
Specifications

Specifications

SORD
Monoclonal
0.57 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q00796
HEL-S-95n, SORD1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6652
Western Blot
OTI13G5
Unconjugated
SORD
Liquid
SORD
Full length human recombit protein of human SORD produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit