Learn More
SORD Mouse anti-Human, Unconjugated, Clone: 4A8E1, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 676251IG150UL
This item is not returnable.
View return policy
Description
The SORD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SORD. This antibody recognizes human antigen. The SORD antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
SORD | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, ELISA | |
4A8E1 | |
Unconjugated | |
150 μL | |
6652 | |
IgG2b |