Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Sortilin Monoclonal Antibody (CL6528)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531438
Description
Immunogen sequence: LERNCEEKDY TIWLAHSTDP EDYEDGCILG YKEQFLRLRK SSVCQNGRDY VVTKQPSICL CSLEDFLCDF GYYRPENDSK CVEQPELKGH DLEFCLYGRE EHLTTNGYRK IPGDKCQGGV NPVREVKD.
Sortilin functions as a sorting receptor in the Golgi compartment and as a clearance receptor on the cell surface. Sortilin is required for protein transport from the Golgi apparatus to the lysosomes by a pathway that is independent of the mannose-6-phosphate receptor (M6PR). It is also required for protein transport from the Golgi apparatus to the endosomes. Sortilin promotes neuronal apoptosis by mediating endocytosis of the proapoptotic precursor forms of BDNF (proBDNF) and NGFB (proNGFB). Also, Sortilin acts as a receptor for neurotensin.
Specifications
Sortilin | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PHU5, Q99523 | |
Sort1 | |
Recombinant protein corresponding to Human SORT1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
CL6528 | |
Unconjugated | |
Sort1 | |
100 kDa NT receptor; 2900053A11Rik; AI852375; Glycoprotein 110; glycoprotein 95; Gp110; Gp95; LDLCQ6; mNTR3; neurotensin receptor 3; NT3; NTR3; Nts3; Ntsr3; Sort1; Sortilin; sortilin 1 | |
Mouse | |
Protein A | |
RUO | |
20661, 6272 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction