Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SNX11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550878
Description
The antibody detects endogenous levels of total Sorting nexin-11 / SNX11 protein.
SNX11 Polyclonal antibody specifically detects SNX11 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
SNX11 | |
Polyclonal | |
Unconjugated | |
SNX11 | |
4933439F10Rik; A930041K09Rik; AI117694; MGC111019; SNX11; sorting nexin 11; sorting nexin-11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29916 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9Y5W9 | |
SNX11 | |
Full length fusion protein of Human SNX11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction