Learn More
Thermo Scientific™ Sorvall™ Adapters for Mach 1.6™ Centrifuges with 4-Place Swing Rotor and Round Buckets
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Manufacturer:  Thermo Scientific™ 75007579
 View more versions of this product
Catalog No. 75007579
Add to cart
Description
Description
For use with four-place swinging bucket rotor 75-002-000 when equipped with round bucket 75-002-002
Adapters sold individually; order correct quantity to equip your rotor.
Specifications
Specifications
7 mL Blood
4-place Swing rotor with round bucket
7 mL