Thermo Scientific™ Sorvall™ CW2 Plus Cell Washer Available on GSA/VA Contract for Federal Government customers only.

Simplify work and save time washing blood cells for antiglobulin reagent tests such as ABO compatibility, Rh testing, cross-matching and the Coombs procedure.

Manufacturer: Thermo Scientific™ 80300567

 View more versions of this product

Catalog No. 22-243-600

  Check Availability
Add to Cart

    Specifications

    Capacity 12 tubes - 12mm x 75mm or 10mm x 75mm tubes
    Length (Metric) Exterior 43.4cm
    Width (Metric) Exterior 32cm
    Electrical Requirements 120/240V 60Hz
    Dimensions (L x W x H) Exterior 17.1 x 12.6 x 24.7 in. (43.4 x 32 x 62.7cm)
    Voltage 120/240V
    Hertz 60Hz
    Length (English) Exterior 17.1 in.
    Net Weight (English) 34 lb.
    Net Weight (Metric) 15.4kg
    Width (English) Exterior 12.6 in.
    Description Sorvall CW2 Plus Cell Washer
    Max. RCF 1200 x G
    Item Description 120/240V 60Hz
    Speed 3550rpm at 60Hz
    Certifications/Compliance 510(k) number BK820020. CSA NRTL, CSA, CE
    Height (English) Exterior 24.7 in.
    Height (Metric) Exterior 62.7cm

    Simplify work and save time in washing blood cells for antiglobulin reagent tests such as ABO compatibility, Rh testing, cross-matching and the Coombs procedure with Thermo Scientific™ Sorvall™ CW2 Plus Cell Washer. The reliable choice, it is designed to support the most common applications requiring the automatic washing of donor blood.

    Fully Automated for Reproducible Results

    • Automated Blood Cell Washing: Standardized, consistent procedures eliminate the variations that result from manual protocols
    • Automatic Alerts: Indicate if the saline level is low, and when a run is complete

    Easy, Reliable Operation

    • Fast, Accurate Results: A time-saving wash cycle, easy to read digital displays and easy-to-use touchpad controls ensure standardized, reproducible procedures
    • Simple to Use: Simply insert the sample tubes, select the test parameters and achieve reliable results in minutes

    Compact and Durable

    • Small Footprint, fits neatly on lab bench or desktop
    • Solid Construction: Low-maintenance design provides years of trouble-free use
    • Brushless motor with sealed-in lubricated bearings
    • Pump tubing is easily changed
    • High-impact, corrosion-free plastic housing

    Ordering Information:

    Replacement Tubing Kit (50019), Replacement Rotor (004770F), and Spare Parts Kit (with tubing, 50008) are available separately.

    Includes:

    Rotor (Product No. 004770F)

    Recommended for:

    Washing and high-speed spin for pelleting red blood cells

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit