Learn More
Thermo Scientific™ Sorvall™ Legend™ Micro 21R Microcentrifuge Available on GSA/VA Contract for Federal Government customers only.
Spin 24 x 2 mL at up to 21,100xg with refrigeration to accelerate routine microcentrifuge tube preparation.
Manufacturer:  Thermo Scientific™ 75002489
 View more versions of this product
Catalog No. 75002489
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications
Description
Ideal for everyday laboratory use up to 21,100xg, Thermo Scientific Sorvall Legend Micro 21R microcentrifuges provide outstanding processing power, unique safety features and a broad array of microcentrifuge rotors and accessories for microcentrifuge tube processing. Refrigeration offers cooling from room temperature to set temperature in just 9 minutes.
Description & Specifications
Specifications
24 x 1.5/2mL
21,100xg
-9° to +40°C
Benchtop centrifuge
Sorvall Legend Micro 21R Centrifuge, Refrigerated
2 years
1 to 99min (1min increments); HOLD
Thermo Scientific Sorvall
14,800rpm
50dbA
CSA, CE, IVD
24 x 1.5/2.0mL rotor with ClickSeal biocontainment lid and Dual Row 18 x 2.0/0.5mL rotor with screw-on lid
230V 50/60Hz
Yes
IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
62.0lb. (28.0kg)
Accessories