Learn More
Selleck Chemical LLCSupplier Diversity Partner SOTALOL, SOTALEX, GRTR99PRCNT DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S2509200MG
Sotalol is a non-selective beta blocker and a potassium channel blocker with an IC50 of 43 μM.
Catalog No. 50-494-6