Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SouthernBiotech™ Genetically Distinct Collagens: Mouse Type IV
Highly purified fibrous protein for a variety of applications
Supplier: SouthernBiotech™ 125004S
Description
Mouse origin, Type IV.
- Can be used as collagen a standard, antigen for antibody production, and coating material in cell culture studies, and in the formation of collagen gels
- Prepared via controlled and limited pepsin digestion; greater than 90% pure
- Supplied as a 0.5mg/mL solution in 500mmol/L acetic acid
Specifications
0.5 mg/mL | |
Controlled and limited pepsin digestion, followed by selective salt precipitation | |
Mouse | |
Store at 2°C to 8°C or in small aliquots at -20°C | |
>90% by SDS-PAGE |
Cell Culture Coating Material, Antigen for Antibody Production, Collagen Gel Formation, Collagen Standard | |
0.5 mg | |
Collagen, Mouse Type IV | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction