Learn More
IgG Human, LE/AF (Low Endotoxin/Azide Free), Polyclonal, Isotype Control, Southern Biotech™
Human Polyclonal Isotype Control
Supplier: Southern Biotechnology Associates 016014
Description
SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.
Specifications
ELISA | |
0.5mg/mL | |
Human | |
RUO | |
Store at -20°C |
Low Endotoxin/Azide-free (LE/AF) | |
Supplied in PBS | |
0.5 mg | |
Primary | |