Learn More
IgG1 Human, Unconjugated, Clone: , Isotype Control, Southern Biotech™
Human Isotype Control
Supplier: Southern Biotechnology Associates 0151K01
Description
SouthernBiotech isotype controls and purified immunoglobulins are useful as negative controls and standards in a variety of immunoassays.
Specifications
ELISA, Flow Cytometry, Immunoprecipitation, In vitro Assay | |
Unconjugated | |
Human | |
RUO | |
Store at 2-8°C | |
IgG1 |
0.5mg/mL | |
Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added | |
0.5 mg | |
Primary | |
Liquid |