Learn More
SouthernBiotech™ Neutralite Avidin
SDP
Neutralized, deglycosylated avidin purified from egg white
Supplier:  SouthernBiotech™ 720004
 View more versions of this product
Catalog No. OB7200-04
Explore available promotions
Add to Cart
Alert:
Reagents contain sodium azide. Sodium azide is very toxic if ingested or inhaled. Avoid contact with skin, eyes, or clothing. Wear eye or face protection when handling. If skin or eye contact occurs, wash with copious amounts of water. If ingested or inhaled, contact a physician immediately. Sodium azide yields toxic hydrazoic acid under acidic conditions. Dilute azide-containing compounds in running water before discarding to avoid accumulation of potentially explosive deposits in lead or copper plumbing.
Description
Description
For Research Use Only. Not for Diagnostic or Therapeutic Use.
Flow cytometry; Enzyme-Linked-Immunosorbent-Assay (ELISA); Immunoblotting; Immunohistochemistry
Order Info
To ensure lot-to-lot consistency, each batch of product is tested by flow cytometry, particle concentration, FLISA or ELISA to conform to characteristics of a standard reference reagent.
Specifications
Specifications
Colorless to Light Brown
8
Egg White
Liquid
d-Biotin
AP
1 mL
2°C to 8°C
ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.