Learn More
SouthernBiotech™ Streptavidin Reagents
SDP
Offer less nonspecific binding than egg white avidin
Supplier:  SouthernBiotech™ 710009M
 View more versions of this product
Catalog No. OB7100-09M
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
0.5 mg/mL
8.2
Streptomyces avidinii
ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry
R-Phycoerythrin
0.5 mg
2°C to 8°C
d-Biotin