Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SouthernBiotech™ Streptavidin Reagents

Offer less nonspecific binding than egg white avidin

$52.60 - $458.50

Specifications

Source Streptomyces avidinii
Specificity d-Biotin
Storage Requirements 2° to 8°C
View More Specs

Products 22
Catalog Number Mfr. No. Conjugate For Use With (Application) pH Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Conjugate For Use With (Application) pH Quantity Price Quantity & Availability  
OB710016L
View Documents
SouthernBiotech™
710016L
R-Phycoerythrin/Cyanine 5 Flow Cytometry, Immunohistochemistry 8.2 0.5mg Each for $453.50
Add to cart
 
OB7100-02S
View Documents
SouthernBiotech™
710002S
Fluorescein Isothiocyanate ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry 8.2 0.5mg Each for $101.10
Add to cart
 
OB7100-03
View Documents
SouthernBiotech™
710003
Rhodamine ELISA, Flow Cytometry, Immunohistochemistry 7.4 1mg Each for $122.00
Add to cart
 
OB710016S
View Documents
SouthernBiotech™
710016S
R-Phycoerythrin/Cyanine 5 Flow Cytometry, Immunohistochemistry 8.2 0.1mg Each for $203.50
Add to cart
 
OB710011L
View Documents
SouthernBiotech™
710011L
Allophycocyanin (APC) Flow Cytometry, Immunohistochemistry 8.2 1mg Each for $553.50
N/A
Due to product restrictions, please to purchase or view availability for this product.
OB710011S
View Documents
SouthernBiotech™
710011S
Allophycocyanin (APC) Flow Cytometry, Immunohistochemistry 8.2 0.1mg Each for $117.10
Add to cart
 
OB710017L
View Documents
SouthernBiotech™
710017L
R-Phycoerythrin/Cyanine 7 Flow Cytometry, Immunohistochemistry 8.2 0.5mg Each for $458.50
Add to cart
 
OB710017S
View Documents
SouthernBiotech™
710017S
R-Phycoerythrin/Cyanine 7 Flow Cytometry, Immunohistochemistry 8.2 0.1mg Each for $187.00
Add to cart
 
OB7100-02
View Documents
SouthernBiotech™
710002
Fluorescein Isothiocyanate ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry 7.4 1mg Each for $178.10
Add to cart
 
OB7100-04
View Documents
SouthernBiotech™
710004
Alkaline Phosphatase ELISA, Immunohistochemistry, Western Blotting 8 1mL Each for $170.00
Add to cart
 
OB7100-07
View Documents
SouthernBiotech™
710007
Texas Red ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry 7.4 1mg Each for $161.10
Add to cart
 
OB7100-06
View Documents
SouthernBiotech™
710006
beta-Galactosidas ELISA, Immunohistochemistry, Western Blotting 7.4 1mL Each for $161.10
Add to cart
 
OB7100-09L
View Documents
SouthernBiotech™
710009L
R-Phycoerythrin ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry 8.2 1mg Each for $390.50
Add to cart
 
OB7100-09S
View Documents
SouthernBiotech™
710009S
R-Phycoerythrin ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry 8.2 0.1mg Each for $52.60
Add to cart
 
OB7100-09M
View Documents
SouthernBiotech™
710009M
R-Phycoerythrin ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry 8.2 0.5mg Each for $241.00
Add to cart
 
OB7100-13S
View Documents
SouthernBiotech™
710013S
Spectral Red Flow Cytometry, Immunohistochemistry 8.2 0.1mg Each for $117.10
Add to cart
 
OB710024
View Documents
SouthernBiotech™
710024
Cyanine 3.5 Flow Cytometry, Immunohistochemistry 7.4 0.1mg Each for $131.10
Add to cart
 
OB710010
View Documents
SouthernBiotech™
710010
R-phycoerythrin-Texas Red ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry 7.4 0.1mg Each for $256.50
Add to cart
 
OB710012
View Documents
SouthernBiotech™
710012
Cyanine 3 ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry 7.4 0.1mg Each for $117.10
Add to cart
 
OB710011M
View Documents
SouthernBiotech™
710011M
Allophycocyanin (APC) Flow Cytometry, Immunohistochemistry 8.2 0.5mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
OB7100-01
View Documents
SouthernBiotech™
710001
Unlabeled ELISA, Flow Cytometry, Immunoblotting, Immunohistochemistry 8.1 5mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
OB7100-05
View Documents
SouthernBiotech™
710005
HRP ELISA, Immunohistochemistry, Western Blotting 7.4 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Specifications

Specifications

Streptomyces avidinii
d-Biotin
2° to 8°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit