Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sox-11 Rat anti-Mouse, Clone: 19B2, MilliporeSigma™

Rat Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen Sox-11
Clone 19B2
Concentration 1 mg/mL
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABE1929100
View Documents
MilliporeSigma
MABE1929100UG
100 μg
Each for $387.00
Add to cart
 
MABE192925
View Documents
MilliporeSigma
MABE192925UG
25 μg
Each for $153.40
Add to cart
 
Description

Description

Sox-11 Monoclonal antibody specifically detects Sox-11 in Mouse samples. It is validated for Immunofluorescence, Immunoprecipitation, Western Blotting.
Specifications

Specifications

Sox-11
1 mg/mL
Unconjugated
Rat
Mouse
GST/His-tagged recombinant fragment corresponding to 113 amino acids from the C-terminal half of mouse Sox-11.
Monoclonal
Protein G purified
19B2
Immunofluorescence, Immunoprecipitation, Western Blot
Purified
RUO
Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG2aκ
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit