Learn More
SOX-9 Rabbit anti-Human,Mouse,Rat, KHL, Clone: 2B10, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB55354X25UL
 View more versions of this product
Catalog No. ZRB55354XMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
SOX-9 Monoclonal antibody specifically detects SOX-9 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.
Specifications
Specifications
SOX-9
2B10
KHL
Purified
SOX9
KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal region of human SOX-9.
∼65 kDa observed; 56.14 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
4 x 25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunohistochemistry, Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_000337
Transcription factor SOX-9
Rabbit
IgG
Protein A Purified
Stem Cell Research
Primary
4X